Learn More
Selleck Chemical LLC KY02111 10mg 1118807-13-8
SDP
Supplier:  Selleck Chemical LLC S709610MG
Encompass_Preferred
KY02111 promotes differentiation of hPSCs to cardiomyocytes by inhibiting Wnt signaling, may act downstream of APC and GSK3?. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4395