Learn More
Medchemexpress LLC HY-102028 2mg Medchemexpress, KY1220 CAS:292168-79-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1020282MG
Encompass_Preferred
Medchemexpress, HY-102028 2mg KY1220 CAS:292168-79-7 KY1220 is a compound that destabilizes both β-catenin and Ras, via targeting the Wnt/β-catenin pathway; with an IC50 of 2.1 μM in HEK293 reporter cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7956