Learn More
Cayman Chemical KYLtrIfluoroacetate salt 10mg
SDP
Supplier:  Cayman Chemical 3691110 MG
Encompass_Preferred
An EphA4 inhibitor (IC50 = 4.22 µM); inhibits EphA4-induced growth cone collapse in embryonic chicken retina explants at 5 µM
Catalog No. 50-291-4906