Learn More
Apexbio Technology LLC Kynurenic acid 492-27-3 1g
SDP
Supplier:  Apexbio Technology LLC B62271000
Encompass_Preferred
Kynurenic acid (CAS 492-27-3) is an endogenous metabolite of the kynurenine pathway and functions as a broad-spectrum antagonist of excitatory amino acid receptors including the NMDA and 7 nicotinic acetylcholine receptors Through these actions kynurenic acid modulates glutamatergic and cholinergic neurotransmission It has been studied for its neuroprotective properties and potential involvement in neurodegenerative and neuropsychiatric disorders Kynurenic acid is currently being evaluated in clinical research such as in the FS2 Safety and Tolerability Study in healthy volunteers (ClinicalTrials gov Identifier NCT02340325) to assess its safety profile
Catalog No. 50-000-89560