Learn More
Medchemexpress LLC Kynurenic acid | 492-27-3 | MFCD00006753 | >98.0% | 189.17 | C10H7NO3 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1008065G
Encompass_Preferred
Kynurenic acid is an endogenous tryptophan metabolite used in research to modulate glutamatergic and cholinergic signaling. It functions as a broad-spectrum antagonist of ionotropic glutamate receptors and α7 nicotinic acetylcholine receptors, and as an agonist of GPR35, making it useful for receptor pharmacology and neuronal signaling studies.
  • Endogenous metabolite with neuroactive properties.
  • Broad-spectrum antagonist of NMDA and other ionotropic glutamate receptors.
  • Antagonist of α7 nicotinic acetylcholine receptors.
  • Agonist activity at GPR35/CXCR8.
  • Soluble in DMSO and basic aqueous solutions; limited water solubility.
  • Stable as a powder when stored at -20°C; follow recommended storage conditions.
Catalog No. 50-002-11183