Learn More
Abcam Kynurenic acid sodium salt, Endogenous ionotropic / nicotinic antagonist, 5G
Supplier:  Abcam AB1202565G
Encompass_Preferred
MW 211.15 Da, Purity >99%. Endogenous antagonist at ionotropic, glycine and α7 nicotinic receptors. Neuroprotective in vivo. Soluble in 1 ml water to give specified mM/ml concentration. Find out more. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4639