Learn More
Medchemexpress LLC Kynureninase (recombinant mouse) | 9024-78-6 | >90.0% | 10 UG

Supplier: Medchemexpress LLC HYP7590410UG
Recombinant mouse kynureninase (full-length) is expressed in Sf9 insect cells with a C-terminal His tag. Supplied as a 0.22 μm filtered solution in 20 mM Tris, 500 mM NaCl, 10% glycerol (pH 8.0), shipped on dry ice, and stored at -80°C. Suitable for enzymatic assays, activity characterization, and biochemical studies.
- Recombinant full-length mouse enzyme (Met1-Ser464).
- C-terminal His tag for purification and detection.
- Expression system: Sf9 insect cells.
- Specific activity >100 pmoles/min/μg (3-hydroxykynurenine substrate).
- Purity >90% by reducing SDS-PAGE.
- Approximate molecular weight 47 kDa.
- Formulated in 20 mM Tris, 500 mM NaCl, 10% glycerol, pH 8.0.
- Endotoxin <1 EU/μg; shipped on dry ice.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.