Learn More
Medchemexpress LLC L-006235 | 294623-49-7 | MFCD18086856 | >=98.0% | 466.60 g/mol | C24H30N6O2S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10335250MG
Encompass_Preferred
L-006235 (Standard) is the analytical reference standard of L-006235, a potent, selective, reversible cathepsin K inhibitor used in research and analytical assays. The standard is formulated for qualitative and quantitative HPLC, GC, and MS applications and supports biochemical and bone-resorption studies.
  • Analytical standard suitable for HPLC, GC, and MS
  • Potent cathepsin K inhibitor with ~5 nM activity in bone resorption assays
  • High selectivity over related cathepsins, reducing off-target activity
  • Shown to reduce collagen degradation and prevent bone loss in assays
  • Available in multiple pack sizes, including 50 mg
  • Chemical formula C24H30N6O2S, molecular weight 466.60 g/mol
Catalog No. 50-002-44831