Learn More
Medchemexpress LLC HY-103352 5mg Medchemexpress, L-006235 CAS:294623-49-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1033525MG
Encompass_Preferred
Medchemexpress, HY-103352 5mg L-006235 CAS:294623-49-7 L-006235 (L-235) is a potent, selective, reversible and orally active inhibitor of cathepsin K, with an IC50 of 5 nM in bone resorption assay. L-006235 shows selectivity for cathepsin K (Ki=0.2 nM) over cathepsin B, cathepsin L, and cathepsin S (Ki=1, 6, and 47 μM, respectively). L-006235 can reduce collagen degradation and prevent bone loss. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0668