Learn More
Medchemexpress LLC L 012 sodium salt | 143556-24-5 | 99.9% | 310.67 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10853710MG
Encompass_Preferred
L 012 sodium salt is a luminal-based chemiluminescent probe designed to detect NADPH oxidase (Nox)-derived superoxide and nitrogen species (reactive oxygen species and reactive nitrogen species).
  • Luminal-based chemiluminescent probe
  • Detects superoxide and nitrogen species
  • Recommended stock solution at 10 mM in DMSO or ultrapure water
  • Store at -20°C or -80°C in dark after aliquoting
  • Avoid repeated freezing and thawing
  • Working solution should be prepared in PBS or serum-free cell culture medium
  • Used in NF-κB-luciferase reporter mice
Catalog No. 50-003-45420