Learn More
Cayman Chemical L-156 602 2.5mg
SDP
Supplier:  Cayman Chemical 238962.5 MG
Encompass_Preferred
A C5a receptor antagonist (IC50 = 1.7 UG/L); an antibiotic active against Gram-positive bacteria; decreases MDP-induced inflammation in mice when administered at a dose of 0.5 mg/kg and concanavalin A-induced inflammation at doses of 0.25 and 0.5 mg/kg
Catalog No. 50-299-4392