Learn More
Enzo Life Sciences L-161-982 (5 mg). CAS: 147776-06-5
SDP
Supplier:  Enzo Life Sciences BMLRA1180005
Encompass_Preferred
A potent and selective prostaglandin EP4 receptor antagonist. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1724