Learn More
TARGETMOL CHEMICALS INC L-168049 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T228935MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. L-168049 is a selective and non-competitive antagonist of human glucagon receptor with IC50s of 3.7 nM 63 nM and 60 nM for human murine and canine respectively. purity: 100%
Catalog No. 50-258-0914