Learn More
Selleck Chemical LLC L 189 S0102-25MG
SDP
Supplier:  Selleck Chemical LLC S010225MG
Encompass_Preferred
L 189 is a novel human DNA ligase inhibitor with IC50 of 5 M 9 M and 5 M for hLigI hLigIII and hLigIV respectively L 189 inhibits base excision repair (BER) and non-homologous end joining (NHEJ) L 189 specifically sensitizes cancer cells to DNA damage
Catalog No. 50-000-39391