Learn More
AdipoGen L-2 4-Diaminobutyric acid 2HCl
SDP
Supplier:  AdipoGen CDXD0154G005
Encompass_Preferred
Chemical. CAS 1883-09-6. Formula C4H10N2O2 . 2HCl. MW 118.1 . 72.9. Synthetic Unnatural amino acid derivative. Pharmacological tool and potential chiral building block used as an internal standard for amino acid analysis. Suitable reagent used for the differentiation of beta-N-methylamino-L-alanine from the diamino acids by using HPLC-FD, UHPLC-UV, UHPLC-MS, and triple quadrupole tandem mass spectrometry UHPLC-MS/MS. Used in the quantification of neurotoxin beta-N-methylamino-L-alanine BMAA in seafood. It has been found to inhibit GABA transaminase ABAT, producing elevated levels of GABA and to have an antitumor activity.
Catalog No. 50-256-1210