Learn More
Medchemexpress LLC L-(+)-2-amino-4-phosphonobutyric acid | 23052-81-5 | >98.0% | 183.1 g/mol | C4H10NO5P | 1 ML
SDP
Supplier:  Medchemexpress LLC HY100781A10MM 1ML
Encompass_Preferred
L-AP4 (also called L-APB) is a selective group III metabotropic glutamate receptor agonist used in neuropharmacology and receptor pharmacology studies. It is supplied as a high-purity research reagent commonly provided as a ready-to-use solution or as solids for assay preparation.
  • Selective agonist for group III mGluRs (mGlu4, mGlu8, mGlu6, mGlu7).
  • Reported EC50 values: mGlu4 0.13 μM; mGlu8 0.29 μM; mGlu6 1.0 μM; mGlu7 249 μM.
  • High purity (>98%).
  • Available as a 10 mM solution (1 mL) or as solid quantities (e.g., 1 mg, 10 mg).
  • Suitable for in vitro pharmacology, electrophysiology, and receptor activation assays.
Catalog No. 50-002-11298