Learn More
Medchemexpress LLC L-2-Aminobutyric acid | 1492-24-6 | 99.4% | 103.12 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0105891G
Encompass_Preferred
L-2-Aminobutyric acid (H-Abu-OH) is a non-proteinogenic amino acid found in human tissues and body fluids. It is formed by the transamination of oxobutyric acid. This compound can be used as a precursor for anticonvulsant and anti-tuberculosis agents and is a crucial intermediate in the chemical and pharmaceutical industries. It is elevated in the plasma of children with various conditions.
  • Found in human tissues and body fluids
  • Precursor for anticonvulsant and anti-tuberculosis agents
  • Crucial intermediate in chemical and pharmaceutical industries
  • Elevated in plasma in certain medical conditions
Catalog No. 50-003-66527