Learn More
Bachem H-b-(2-Thienyl)-Ala-OH
SDP
Supplier:  Bachem 4006811.0005
Encompass_Preferred
H-b-(2-Thienyl)-Ala-OH, 22951-96-8, C7H9NO2S, Mr 171.22, 5g. L-b-(2-Thienyl)-alanine (H-Thi-OH) is structurally similar to L-phenylalanine. H-Thi-OH inhibits the growth of Escherichia coli, presumably by competitive inhibition of enzymes for which Phe is the substrate.
Catalog No. 50-194-6680