Learn More
Medchemexpress LLC L-368,899 hydrochloride | 160312-62-9 | 100.0% | 591.23 | 5 MG
Supplier: Medchemexpress LLC 5MGHY108677
L-368,899 hydrochloride is a potent, selective, orally bioavailable, non-peptide oxytocin receptor antagonist. It functions as a tocolytic agent, showing high affinity for oxytocin receptors in both rat and human uterus.
- Potent and selective oxytocin receptor antagonist
- Orally bioavailable compound
- Exhibits IC50s of 8.9 nM for rat uterus oxytocin receptor and 26 nM for human uterus oxytocin receptor
- Shows less activity on VP receptor in human and rat liver and kidney