Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-371,257 | 162042-44-6 | 99.3% | 507.58 | 10 MM / 1 ML
SDP

Supplier:  Medchemexpress LLC HY1501010MM/1ML

Encompass_Preferred

L-371,257 is an orally bioavailable, non-blood-brain barrier penetrant, selective, and competitive antagonist of the oxytocin receptor (pA2=8.4). It exhibits high affinity at both the oxytocin receptor (Ki=19 nM) and vasopressin V1a receptor (Ki=3.7 nM).

  • Orally bioavailable
  • Non-blood-brain barrier penetrant
  • Selective and competitive antagonist of the oxytocin receptor
  • High affinity at oxytocin receptor (Ki=19 nM)
  • High affinity at vasopressin V1a receptor (Ki=3.7 nM)
  • Appears as a white to off-white solid
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at 8.33 mg/mL (16.41 mM)

Catalog No. 50-003-60490


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.