Learn More
Medchemexpress LLC L-4-Chlorophenylglycine | 67336-19-0 | 185.61 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0057205G
Encompass_Preferred
L-4-Chlorophenylglycine, also known as H-Phg(4-Cl)-OH, is a Glycine derivative and an amino acid derivative with a molecular formula of C8H8ClNO2 and a molecular weight of 185.61. This high-purity compound, typically at 98.85%, is commonly used in various research and synthesis applications.
  • Amino acid derivative
  • High purity (98.85%)
  • Precisely defined molecular structure (C8H8ClNO2)
  • Consistent molecular weight (185.61)
  • Suitable for research and synthesis
  • Appears as a solid
  • White to off-white in color
  • Soluble in DMSO and water (with specific conditions)
Catalog No. 50-004-49312