Learn More
Medchemexpress LLC L-5,6,7,8-tetrahydrofolic acid | 135-16-0 | 96.8% | C19H23N7O6 | 10MG
SDP
Supplier:  Medchemexpress LLC HY1452010MG
Encompass_Preferred
Tetrahydrofolic acid (L-5,6,7,8-tetrahydrofolic acid) is a biologically active folate derivative that functions as a donor of one-carbon units in amino acid, nucleic acid, and lipid metabolism. It is used in biochemical and pharmaceutical research involving folate-mediated one-carbon transfer. The compound is unstable in solution and is stored frozen, protected from light; DMSO is a suitable solvent.
Catalog No. 50-002-60805