Learn More
Medchemexpress LLC L-5-Hydroxytryptophan | 4350-09-8 | 99.99% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYB1716
Encompass_Preferred
L-5-Hydroxytryptophan (L-5-HTP), a naturally occurring amino acid and dietary supplement, is used as an antidepressant, appetite suppressant, and sleep aid. It serves as the immediate precursor of the neurotransmitter serotonin and acts as a reserpine antagonist. It is also utilized in the treatment of fibromyalgia, myoclonus, migraine, and cerebellar ataxia.
  • Endogenous metabolite
  • Naturally occurring amino acid
  • Dietary supplement
  • Precursor of neurotransmitter serotonin
  • Reserpine antagonist
  • Used for research purposes only; not sold to patients
Catalog No. 50-004-13918