Learn More
Medchemexpress LLC L 674573 | 127481-29-2 | 99.5% | 5 MG

Supplier: Medchemexpress LLC HY1205715MG
L 674573 (CAS 127481-29-2) is a small-molecule leukotriene biosynthesis inhibitor used for preclinical research. Supplied as a high-purity solid and intended for laboratory assays; for research use only.
- Inhibits leukotriene biosynthesis
- Molecular formula C28H27NO3S, molecular weight ≈ 457.6 g·mol^-1
- High purity with certificate of analysis available
- Supplied as a powder; store at -20°C
- For research use only, not for human or veterinary use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.