Learn More
Medchemexpress LLC L-690330 hydrate | 2930564-26-2 | 98.0% | 316.14 g/mol | C8H14O9P2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY101075A5MG
Encompass_Preferred
L-690330 hydrate is a research-grade small-molecule inhibitor of inositol monophosphatase (IMPase), supplied for biochemical and cell-based studies to probe IMPase activity and downstream signaling. The compound is provided as solids and ready-to-use DMSO solutions for immediate use in assays.
  • Competitive inhibitor of inositol monophosphatase with low micromolar Ki values.
  • High purity (≥98.0%) appropriate for research applications.
  • Available as small solid quantities and pre-made DMSO solution for convenience.
  • Molecular weight 316.14 g/mol; formula C8H14O9P2.
  • Demonstrated to induce P-AMPK and autophagy in cell-based assays.
  • Supporting documents (datasheet, COA, SDS) available.
Catalog No. 50-002-13394