Learn More
Medchemexpress LLC HY-10957 5mg Medchemexpress, L-692429 CAS:145455-23-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY109575MG
Encompass_Preferred
Medchemexpress, HY-10957 5mg L-692429 CAS:145455-23-8 L-692429 (MK-0751) is a benzolactam derivative and a nonpeptidyl growth hormone secretagogue (GHS) agonist. L-692429 binds to G protein-coupled receptor with a Ki of 63 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0772