Learn More
Medchemexpress LLC L-692429 (MK-0751) | 145455-23-8 | 99.9% | 509.60 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10957100MG
Encompass_Preferred
L-692429 (MK-0751) is a benzolactam derivative and a nonpeptidyl growth hormone secretagogue (GHS) agonist. It binds to G protein-coupled receptor with a Ki of 63 nM. This product is for research use only.
  • Stimulates intracellular calcium release, inositol phosphate (IP) turnover, cAMP-responsive element binding protein (CREB) activity, serum-responsive element activity, and bioluminescence resonance energy transfer (BRET) activity.
  • Increases fluorescent emission in HeLa-T4 cells transiently expressing the flag epitope-tagged growth hormone secretagogue (GHS) receptor.
  • Activates the GHS receptor, significantly increasing luciferase activity and suggesting stimulation of the MAPK pathway.
Catalog No. 50-003-44154