Learn More
Medchemexpress LLC L-692429 (mk-0751) | 145455-23-8 | 99.9% | 509.60 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1095710MM/1ML
Encompass_Preferred
L-692429 (MK-0751) is a benzolactam derivative and a nonpeptidyl growth hormone secretagogue (GHS) agonist that binds to G protein-coupled receptors with a Ki of 63 nM. This product is for research use only. It stimulates intracellular calcium release, inositol phosphate (IP) turnover, cAMP-responsive element binding protein (CREB) activity, serum-responsive element activity, and bioluminescence resonance energy transfer (BRET) activity. L-756867 inhibits L-692429-stimulated GH secretion in anesthetized rats.
  • Binds to G protein-coupled receptors
  • Stimulates intracellular calcium release (EC50: 26 nM)
  • Stimulates IP turnover (EC50: 47 nM)
  • Stimulates CREB activity (EC50: 60 nM)
  • Stimulates SRE activity (EC50: 63 nM)
  • Stimulates BRET activity (EC50: 58 nM)
  • Activates MAPK pathway in hela-T4 cells
  • Inhibits GH secretion in anesthetized wistar rats
Catalog No. 50-003-45426