Learn More
Medchemexpress LLC L-742001 hydrochloride | 174605-64-2 | MFCD22587878 | 99.0% | 450.35 g·mol⁻1 | C23H25Cl2NO4 | 10 MG

Supplier: Medchemexpress LLC HY11836810MG
L-742001 hydrochloride is a research reagent that inhibits the influenza virus PA endonuclease, with an observed EC90 of 4.3 μM for vRNP activity in HEK293T cells. It is supplied as a white to off-white solid with reported high purity and is intended for laboratory research use only.
- CAS number 174605-64-2.
- Molecular formula C23H25Cl2NO4.
- Molecular weight 450.35 g·mol⁻1.
- Purity 99.0%.
- Appearance white to off-white solid.
- Solubility DMSO: 20 mg/mL (44.41 mM); warming and sonication may be required.
- Storage powder: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (6 months) or -20°C (1 month).
- For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.