Learn More
Selleck Chemical LLC L-778123 hydrochloride E4620-100MG
SDP
Supplier:  Selleck Chemical LLC E4620100MG
Encompass_Preferred
L-778123 hydrochloride is a potent dual inhibitor of farnesyl protein transferase (FPTase) with an IC50 of 2 nM and geranylgeranyl protein transferase I (GGPTase-I) with an IC50 of 98 nM It also displays promising anti-leukemia activity
Catalog No. 50-000-39048