Learn More
Medchemexpress LLC L-798106 | 244101-02-8 | C27H22BrNO4S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1527410MM/1ML
Encompass_Preferred
L-798106, also known as CM9 or GW671021, is a highly potent and selective antagonist for the prostanoid EP3 receptor, exhibiting a Ki value of 0.3 nM. This compound has demonstrated the ability to suppress systemic insulin resistance and inflammation in adipose tissue in animal models.
  • Selective EP3 receptor antagonist.
  • Suppresses systemic insulin resistance and adipose tissue inflammation.
  • Inhibits contractile responses in guinea-pig vas deferens.
  • Modulates ACh release in guinea-pig tracheal smooth muscle.
  • Potential applications in research related to obesity-induced metabolic disorders.
Catalog No. 50-003-73095