Learn More
Medchemexpress LLC L-826266 | 244101-03-9 | 98.0% | 570.88 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY193615MG
Encompass_Preferred
L-826266 is a selective and competitive EP3 receptor antagonist that can be used for convulsive disorders research. It prevents the PGE2-induced decrease of Na+,K+-ATPase activity in adult rat hippocampal slices. In vivo, it delays seizures and increases the latency for clonic and generalized tonic-clonic seizures induced by PTZ.
  • Prevents PGE2-induced decrease of Na+,K+-ATPase activity
  • Delays seizures
  • Increases latency for clonic and generalized tonic-clonic seizures
Catalog No. 50-003-84957