Learn More
Enzo Life Sciences L-858051 (5mg)
SDP
Supplier:  Enzo Life Sciences BMLCN1050005
Encompass_Preferred
A water-soluble forskolin derivative, retaining adenylate cyclase stimulatory activity (EC50=10 µM). Purity: ≥92% (HPLC). Solubility: Soluble in water (>10 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0437