Learn More
Medchemexpress LLC L-888607 (racemate) | 1030017-51-6 | 99.8% | 375.84 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY111271A
Encompass_Preferred
L 888607 Racemate is a selective prostaglandin D2 receptor subtype 1 (DP1) antagonist. It exhibits Ki values of 132 nM for DP1 and 17 nM for thromboxane A2 receptor (TP).
  • Selective prostaglandin D2 receptor subtype 1 (DP1) antagonist.
  • Ki values of 132 nM for DP1 and 17 nM for thromboxane A2 receptor (TP).
  • For research use only.
Catalog No. 50-003-46840