Learn More
Sigma Aldrich Fine Chemicals Biosciences L-α-Lysophosphatidylcholine from egg yolk, 9008-30-4, MFCD00131444, 25mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L412925MG
Encompass
≥99%, Type I, powder, Synonym: 1-Acyl-sn-glycero-3-phosphocholine, L-α-Lysolecithin.
Catalog No. NC1595906