Learn More
Cayman Chemical L-a-Methyldopa 5g
SDP
Supplier:  Cayman Chemical 218145 G
Encompass_Preferred
A DOPA decarboxylase inhibitor (ED50 = 21.8 mg/kg); α2-adrenergic receptor agonist; metabolized to α-methylnorepinephrine in the CNS; has antihypertensive activity in vitro and in vivo; improves trophoblast and endothelial cellular interaction in vitro
Catalog No. 50-298-5840