Learn More
Apexbio Technology LLC L-Acetylcarnitine (hydrochloride) 5080-50-2 5g
SDP
Supplier:  Apexbio Technology LLC C30985000
Encompass_Preferred
L-Acetylcarnitine (hydrochloride) (CAS 5080-50-2) an acetylated derivative of L-carnitine facilitates the mitochondrial uptake of acetyl-CoA supporting fatty acid oxidation and energy metabolism It enhances acetylcholine synthesis and contributes to membrane phospholipid and protein production In vitro L-acetylcarnitine reduces SKOV-3 ovarian cancer cell proliferation at higher concentrations without affecting CA-125 or nerve growth factor receptor expression In vivo studies demonstrate its rapid and sustained antidepressant-like activity in rodent models linked to epigenetic upregulation of mGlu2 (Grm2) receptors L-acetylcarnitine is utilized in research focusing on metabolism neurobiology and psychiatric disorders
Catalog No. 50-000-88069