Learn More
Medchemexpress LLC L-Alanine-1,2,3-13C3, N-[(9H-fluoren-9-ylmethoxy) carbonyl]- | 765259-05-0 | 98.5% | 314.31 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW009204S510MG
Encompass_Preferred
Fmoc-Ala-OH-13C3 is a 13C-labeled alanine derivative. It is intended for research use only and can be utilized as a tracer or an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis
  • Suitable for NMR analysis
  • Suitable for GC-MS analysis
  • Suitable for LC-MS analysis
Catalog No. 50-003-66875