Learn More
Medchemexpress LLC L-Alanine-1,2,3-13C3, N-[(9H-fluoren-9-ylmethoxy)carbonyl]- | 765259-05-0 | 98.5% | 314.31 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW009204S550MG
Encompass_Preferred
Fmoc-Ala-OH-13C3 is a 13C-labeled alanine derivative, intended for research use only. This compound can be utilized as a tracer and an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been integrated into drug molecules, primarily to serve as tracers for quantitation during the drug development process.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, or LC-MS applications.
  • Incorporates stable heavy isotopes.
Catalog No. 50-004-53152