Learn More
Medchemexpress LLC L-alanine-13C3, 15N | 202407-38-3 | MFCD19443802 | 98.0% | 93.06 g·mol⁻1 | 13C3H7 15NO2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0229S1010MG
Encompass_Preferred
L-alanine-13C3,15N is an isotopically labeled form of the amino acid L-alanine in which three carbon atoms are 13C and the nitrogen is 15N. Supplied as a 10 mg vial for research use, it serves as a stable isotope internal standard, tracer, or reference material for analytical and biochemical workflows. Purity is 98.0% and molecular weight is 93.06 g·mol⁻1.
  • Isotopically labeled standard for mass spectrometry and metabolomics.
  • Supplied in a small, research-scale package suitable for analytical assays.
  • High purity suitable for quantitative internal standard applications.
  • Stable, non-radioactive tracer for metabolic and pathway studies.
  • Provided with batch-specific datasheet and certificate of analysis for traceability.
Catalog No. 50-002-96322