Learn More
Medchemexpress LLC L-Alanine-15N, N-[(9H-fluoren-9-ylmethoxy)carbonyl]- | 117398-49-9 | 99% | 312.33 | 50 MG

Supplier: Medchemexpress LLC HYW009204S50MG
Fmoc-Ala-OH-15N is an isotopic labeled compound, specifically 15N labeled Fmoc-Ala-OH. Stable heavy isotopes of elements like hydrogen, carbon, and nitrogen are incorporated into drug molecules to serve as tracers for quantitation during drug development. Such labeling can influence the pharmacokinetic and metabolic profiles of drugs.
- Isotopic labeled compound
- Purity of 99% (HPLC)
- Isotopic enrichment of 99%
- Functions as a tracer for drug development
- Influences pharmacokinetic and metabolic profiles of drugs
- Appears as a white to off-white solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.