Learn More
Medchemexpress LLC L-alanine-2,3,3,3-d4 | 18806-29-6 | MFCD00084109 | 99.9% | 93.12 | C3H3D4NO2 | 10 MG

Supplier: Medchemexpress LLC HYN0229S310MG
L-Alanine-d4 is the deuterium-labeled form of L-alanine supplied as a high-purity stable isotope reagent for analytical and biochemical applications. Provided as a 10 mg solid, it is suitable for metabolic tracing, mass spectrometry internal standards, and isotope-dilution quantitation.
- Deuterated L-alanine: 2,3,3,3-d4.
- High purity: 99.9%.
- Molecular formula: C3H3D4NO2; molecular weight: 93.12 g/mol.
- Package size: 10 mg solid.
- Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.