Learn More
Chem-Impex International, Inc. L-Alanine-2-chlorotrityl resin (0.3 - 1.0 meq/g, 200 - 400 mesh) | MFCD00801399 | 5G
Supplier:  Chem-Impex International, Inc. 034495G
Encompass_Preferred
L-Alanine-2-chlorotrityl resin (0.3 - 1.0 meq/g, 200 - 400 mesh), MFCD00801399, 5G
Catalog No. 50-497-879