Learn More
Medchemexpress LLC L-alanine-2-d1 | 21386-65-2 | MFCD00084105 | >=98.0% | 90.10 g/mol | C3H6DNO2 | 500 MG

Supplier: Medchemexpress LLC HYW141857500MG
L-Alanine-2-d1 is a deuterium-labeled analogue of L-alanine supplied for laboratory research. It is used as a stable isotope tracer and as an internal standard to improve accuracy and quantitation in analytical workflows such as NMR, GC-MS, and LC-MS.
- Deuterium-labeled tracer for metabolic and pharmacokinetic studies.
- Suitable as internal standard for quantitative NMR, GC-MS, and LC-MS analyses.
- White to off-white solid powder for laboratory handling.
- High chemical purity (≥98%) for analytical reliability.
- Stable under cold storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
- Molecular formula C3H6DNO2; molecular weight ~90.10 g/mol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.