Learn More
Medchemexpress LLC L-Alanine-3-13C-2-d | 160033-81-8 | 98.0% | 91.09 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0229S4
Encompass_Preferred
L-Alanine-13C,d is the 13C- and deuterium labeled L-Alanine. L-Alanine is a non-essential amino acid, involved in sugar and acid metabolism, increases immunity, and provides energy for muscle tissue, brain, and central nervous system.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-30670