Learn More
Medchemexpress LLC L-alanine-3-13C | 65163-25-9 | 99.0% | 10 MG

Supplier: Medchemexpress LLC HYN0229S610MG
L-Alanine-3-13C is a 13C-isotopically labeled form of L-alanine intended for use as an isotopic tracer or internal standard in metabolic and mass-spectrometry studies. It is supplied at high purity for analytical applications and is provided with supporting documentation for quality and safety.
- 13C label at the 3-position for isotopic tracing
- High purity (99.0%) suitable for analytical workflows
- Available in small laboratory quantities (10 MG, 25 MG, 50 MG, 100 MG)
- Powder storage: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (6 months), -20°C (1 month)
- Includes product data sheet, certificate of analysis, and safety data sheet
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.