Learn More
Medchemexpress LLC L-alanine, 3-azido-, hydrochloride | 1620171-64-3 | MFCD18382048 | 98.0% | 166.57 g/mol | C3H7ClN4O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW33632825MG
Encompass_Preferred
3-Azido-L-alanine hydrochloride is an azide-functionalized amino acid supplied as a solid hydrochloride salt for research use. It provides an azide handle for site-specific bioorthogonal conjugation and is used in biochemical assays and chemical synthesis involving click chemistry.
  • Azide-functionalized amino acid for click chemistry applications.
  • Suitable for copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted cycloaddition (SPAAC).
  • Off-white to pink solid with specified purity.
  • Purity typically reported around 98% (batch-dependent).
  • Store sealed at 4°C; in solution store at -20°C to -80°C for long-term storage.
  • Provided in small quantities for laboratory synthesis and labeling workflows.
Catalog No. 50-002-82183