Learn More
Medchemexpress LLC L-alanine, 3-azido-, hydrochloride | 1620171-64-3 | MFCD18382048 | 98.0% | 166.57 g/mol | C3H7ClN4O2 | 5 MG

Supplier: Medchemexpress LLC HYW3363285MG
3-Azido-L-alanine hydrochloride is an aliphatic, azide-containing amino acid used as a bioorthogonal reagent for labeling and conjugation via click chemistry. Supplied as the hydrochloride salt in solid form, it is suitable for protein and peptide modification using copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC).
- Contains an azide functional group for click chemistry reactions.
- Compatible with CuAAC and SPAAC bioorthogonal labeling.
- Hydrochloride salt, solid form for stability and handling.
- High purity: 98.0%.
- Molecular formula C3H7ClN4O2; molecular weight 166.57 g/mol.
- CAS number 1620171-64-3 for unambiguous identification.
- Available in small research pack sizes suitable for labeling workflows.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.