Learn More
Medchemexpress LLC L-Alanine-d4 (L-2-Aminopropionic acid-d4) | 18806-29-6 | 99.9% | 93.12 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0229S350MG
Encompass_Preferred
L-Alanine-d4 is the deuterium labeled L-Alanine. L-Alanine is a non-essential amino acid, involved in sugar and acid metabolism, increases immunity, and provides energy for muscle tissue, brain, and central nervous system.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-27701