Learn More
Medchemexpress LLC L-Alanine, L-alanyl- | 1948-31-8 | 98.0% | 160.17 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0101621G
Encompass_Preferred
H-Ala-Ala-OH (L-Alanyl-L-alanine; Ala-Ala) is a nonpolar dipeptide absorbed by human intestinal Caco-2 cells. The transport of alanine (Ala), similar to proton/amino acid symport, can lead to cytoplasmic acidification. The product appears as a white to off-white solid.
  • Solid appearance
  • White to off-white color
  • Protect from light
  • Store under nitrogen
Catalog No. 50-004-52461